Orders LinkContactUsLinkComponent
Glo Germ
Glo Germ
Supplier:   GLO GERM SE
Glo Germ
Supplier:  GLO GERM SE
Certificates
Glo Germ
4 Options Available Below
Product Details & Documents
Used Worldwide to Teach Proper Handwashing Technique.
  • Choose Powder, Liquid, or Gel
  • Test Handwashing or Cleaning Effectivity
  • Invisible Until Exposed to UV Light
Both the oil and the powder contain minute pieces of plastic that glow under UV light. To become germ detectives, students apply a small amount of oil before handwashing and then try to spot remaining “germs” under a UV light. To investigate the thoroughness of surface cleaning, students wipe away a light dusting of Glo Germ powder and hunt for germs with the UV light.
Glo Germ gel is recommended where stains are of concern. The adjustable flow Glo Brush lets users apply a thin layer of powder to any surface quickly and without waste and features a refillable powder reservoir with a cover to protect bristles.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 470004-262
    Glo Germ Powder
    Glo Germ Powder
    112 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $533.01
    $533.01
    Each (1UN)
       $533.01
    Supplier Number: POWDER
  • 470004-260
    Glo Germ Liquid
    Glo Germ Liquid
    224 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $572.99
    $572.99
    Each (1UN)
       $572.99
    Supplier Number: OIL
  • 470100-620
    Glo Germ Gel - White
    Glo Germ Gel - White
    224 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $572.99
    $572.99
    Each (1UN)
       $572.99
    Supplier Number: GEL
  • 470100-682
    Glo Germ Brush And Powder
    Glo Germ Brush And Powder
    112 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,559.05
    $1,559.05
    Each (1UN)
       $1,559.05
    Supplier Number: BRHP