Glass Thermometers
Glass Thermometers, -40...70 ºC (-40...160 ºF)
Catalog # 470105-948
Supplier: SPER SCIENTIFIC LTD SE
Número de proveedor #: 738730
Jump to:
Product Details & Documents
All filled with safe red alcohol, white-backed, total immersion, 6" (153 mm) long, in blue plastic holder.
- Pack of 12
- Pocket thermometer
Especificaciones
- Description:Glass Thermometers
- Range:-40...70 ºC (-40...160 ºF)



