Glass Stirring Rods
Rod, Stirring, Glass, 30 cm (12")
Catalog # 470007-284
Supplier: UNITED SCIENTIFIC SUPPLIES
: GLROD12
Jump to:
Product Details & Documents
Fire Polished Flint Glass.
- One rounded end
- One flat end
- Length: 12"" (30 cm)
Flint glass stir rods are non-reactive and perfect for stirring a variety of solutions.
Especificaciones
- Description:Rod, Stirring, Glass
- Length:30 cm (12")