Orders LinkContactUsLinkComponent
Glass Plates
Glass Plates
Supplier:   VWR International
Glass Plates
Supplier:  VWR International
Certificates
Glass Plates
3 Options Available Below
Product Details & Documents
Plates made of soda lime glass, 2mm thick, with ground polished edges. Useful for a variety of general laboratory applications including as a cover, mixing surface, etc.
  • Soda lime glass
  • Polished edges
Clean cut edges, not ground. Available in Clear or Cobalt Blue.
Product Family Options
Información del producto
  • Availability
    Price
  • 470123-224
    Glass Plates, Type: Clear, Size: 2" x 2"
    Glass Plates, Type: Clear, Size: 2" x 2"
    Discontinued
  • 470123-226
    Glass Plates, Type: Clear, Size: 3" x 3"
    Glass Plates, Type: Clear, Size: 3" x 3"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $300.60
    $300.60
    Paquete de 12
       $300.60
  • 470123-228
    Glass Plates, Type: Clear, Size: 4" x 4"
    Glass Plates, Type: Clear, Size: 4" x 4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $338.99
    $338.99
    Paquete de 12
       $338.99
  • 470123-230
    Glass Plates, Type: Clear, Size: 6" x 6"
    Glass Plates, Type: Clear, Size: 6" x 6"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $473.32
    $473.32
    Paquete de 12
       $473.32