Orders LinkContactUsLinkComponent
Glass Depression Slides
Glass Depression Slides
Glass Depression Slides
Certificates
Glass Depression Slides
Glass Depression Slides
3 Options Available Below
Product Details & Documents
These depression slides are ideal for viewing living specimens suspended in liquid.
  • Made with quality sheet glass with ground edges
  • Sold in boxes of 12 slides
  • Available with single or double concavities
These precleaned slides have a round, shallow depression in the center of the slide, making it easy to view materials in liquid.
Cavities measure approximately 16mm in diameter, and 0.8mm deep.
Product Family Options
Single Cavity Slides
Información del producto
  • Thickness
    L×W
    Availability
    Price
  • 470235-728
    Slides, 1 shallow depression
    Slides, 1 shallow depression
    1.0 - 1.2 mm
    3×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $186.50
    $186.50
    Log in Para disponibilidad
    Case de 12 (12UN)
    Case de 12 (12UN) - $203.28
    $203.28
    Paquete de 12
       $186.50
    Case de 12 (12UN)
       $203.28
    Supplier Number: 4-13057-DZ-12
  • 470019-022
    Single concavity slides
    Single concavity slides
    3 mm
    3×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $216.95
    $216.95
    Paquete de 12
       $216.95
    Supplier Number: CSTK01
Double Cavity Slides
Información del producto
  • Thickness
    L×W
    Availability
    Price
  • 470200-930
    Double cavity slides
    Double cavity slides
    1.3 mm
    3×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $170.04
    $170.04
    Paquete de 12
       $170.04
    Supplier Number: CS3X12-PK/12