Glass Cylindrical Mirrors
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Acerca de este artículo
Demonstrate converging and diverging mirrors.
- Self standing
- Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
2 Options Available Below
Product Details & Documents
Demonstrate converging and diverging mirrors.
- Self standing
- Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
Product Family Options
Información del producto
- AvailabilityPrice
- 470007-092Concave/cylindrical mirrorSupplier Number: MRCC01
- 470007-094Convex/cylindrical mirrorSupplier Number: MRCV02