Generalized Animal Cell Slide
Generalized Animal Cell Slide, Section, Hematoxylin and Eosin
Catalog # 470177-556
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates animal cell characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal tissue. Nucleus, nucleolus, chromatin, and cell membrane are illustrated. Study guide included.
Especificaciones
- Description:Generalized Animal Cell Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section