General Connective Tissues Slide Set
Expanded Set Explores Cells and Extracellular Material
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are sections unless otherwise specified.
: Slide Set contents include: Mesenchyme, embryonic; Mucous, umbilical cord; Areolar, fascia (Whole mount); Adipose, mesentery (sect/Whole mount); Chondroid; Adipose; Reticular; Collagenous, rat tail (Whole mount); White Fibrous, tendon; Yellow elastic, ligament (Whole mount); Yellow elastic, ligamentum nuchae; Hyaline cartilage, xiphisternum; Hyaline cartilage, trachea; Elastic, hyaline cartilage, external ear; Elastic cartilage, epiglottis; White fibro-cartilage, invertebral disc; Osteogenesis, intramembranous ossification; Osteogenesis, endochondral ossification; Bone, compact (cs); Bone, compact (cs/ls); Bone development, femur (cs); Bone, ground preparation (cs/ls); Mouse tail (cs); Osteogenesis and ligament development (ls).
- Type:Animal
- Description:General Connective Tissues Slide Set
- No. of Slides:25