Orders LinkContactUsLinkComponent
Galena
Galena
Supplier:   Avantor
Galena
Supplier:  Avantor
Certificates
Galena
3 Options Available Below
Product Details & Documents
Bright metallic cubic cleavages; lead ore.
  • Collected in Morocco
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Specimen Dimensions
    Availability
    Price
  • 470025-584
    Galena, student specimens
    Galena, student specimens
    1×1" - 1×1"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $560.13
    $560.13
    Paquete de 10
       $560.13
  • 470025-588
    Galena, hand specimen
    Galena, hand specimen
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $536.08
    $536.08
    Each (1UN)
       $536.08
  • 470025-586
    Galena bulk pack, 1 kg
    Galena bulk pack, 1 kg
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $591.63
    $591.63
    Each (1UN)
       $591.63