Funnel Support
Funnel Support, Length 16"
Catalog # 470020-354
Supplier: HUMBOLDT MFG CO
Número de proveedor #: H-21650
Jump to:
Product Details & Documents
Featuring slotted holes for four funnels, it is constructed of hardwood with a metal clamp and thumbscrew for attaching it to a support rod.
Especificaciones
- Description:Funnel Support, Length 16"