Fundamentals of Physics Reflection/Refraction Apparatus
Catalog # 470014-332
Supplier: VWR International
Specifications
- Descripción:FUNdamentals of Physics Reflection/Refraction Apparatus
- No. of Lab Groups:1
- Referencia:470014-332
- Nº de proveedor:RRKT
Specifications
About this item
Hands-on investigation even explores light separation.
- 360 degree protractor
- Laser and light source
- Includes teacher manual
Using a plane mirror in addition to a concave and convex mirror with different focal lengths, students encounter reflection through a series of activities. Students also investigate refraction of concave and convex lenses, a Lucite block, and glass block. The base has a 12 V light bulb for investigating light dispersion and also a laser pointer for refraction. The included prism is utilized to demonstrate separation of light into its component colors. Comes with an instruction manual. Required but not included is a 12 V power supply.