Especificaciones
- Descripción:Fundamentals of physics rotational motion apparatus
- Referencia:470014-324
- Nº de proveedor:FN RO-MT
Especificaciones
Acerca de este artículo
Students investigate the relationship between linear and angular velocity.
Apparatus features a 40 cm wide disc with four moveable pegs
- Activity guide included
- Student-friendly process
- Purpose for students: to understand the relationship between linear and angular velocity
Easy to operate, this student-friendly apparatus consists of a 40 cm wide disc driven by a motorized wheel. The disc’s moveable pegs can be positioned at a specific distance from the center, and a photogate reads the frequency of the pegs when placed at different positions on the spinning disc.
When students measure the frequency of the pegs at different positions but a constant rotation, they find the angular velocity to be consistent while the linear velocities are different. The drive motor can be positioned near the center of the disk, causing a faster rotation, or near the edge, which causes a slower rotation.
The fundamentals of physics photogate system and a low voltage power supply (0 - 9 VDC) are required, but available separately.