Fundamentals of Physics Optics Bench
Catálogo # 470014-334
Proveedor: VWR International
Especificaciones
- Descripción:FUNdamentals of Physics Optics Bench
- No. of Lab Groups:1
- Referencia:470014-334
- Nº de proveedor:FN-OPTKIT
Especificaciones
Acerca de este artículo
Convert the fun stand into an optical bench.
- Large colored rods
- Includes teacher manual
Offer your students a hands-on experience with geometric optics, demonstrating polarization, color mixing, and more. The unique design of this device uses supports that attach to the Fun Stand as a horizontal bar and create a student-friendly optical bench for many experiments. Instructions for all activities are also supplied.
Información de accesorios:
Requires, the Fun Stand, available separately. Requires a 12 V power supply, not included.
Entrega:
Kit Includes: (4) color filters; (2) polarizing lenses; support brackets; light source; support stand; and (2) each of concave and convex lenses, and mirrors; all securely organized in a sturdy storage container.