Ward's® Fulgurite
Fulgurite, 1½ - 2"
Catalog # 470025-900
Supplier: Avantor
Jump to:
Product Details & Documents
Unusual glassy tubes of fused sand produced by lightning strikes in desert sediments. Average size: 1.5"- 2".
Especificaciones
- Description:Fulgurite
- Specimen Dimensions:1½ - 2"