Beginners Readings in Science
Catalog # 470047-886
Supplier: HOUGHTON MIFFLIN HARCOURT (HMH) SE
Specifications
- Descripción:Book, From Tadpole to Frog, (Wendy Pfeffer)
- Longitud:19 cm
- Referencia:470047-886
- No. of Pages:32
- Anchura:23 cm
- Nº de proveedor:0064451239
Specifications
About this item
A collection of classic books to introduce young readers to science.
Learn about life cycles, the rain forest, and why things happen. Various grade levels from pre K to 3.