Orders LinkContactUsLinkComponent
Ward's® Frog Models
Ward's® Frog Models
Supplier:   Avantor
Ward's® Frog Models
Supplier:  Avantor
Certificates
Ward's® Frog Models
Ward's® Frog Models
2 Options Available Below
Product Details & Documents
The Most Realistic Frog Model Available
  • Highly Durable Rubberized Material
  • Common Dissection Organs Highlighted
  • Easily Cleaned And Stored
The meticulous detail and incredible realism of both life-size Rana catesbiana models are extraordinary; even the structures of the inner mouth are shown. Cast from actual specimens, they feature over 50 details from the digestive, circulatory, musculatory, and reproductive systems, colorfully yet subtly painted to realistically show all major structures. Each is made of a unique material for flexibility and durability.
Documents
Product Family Options
Información del producto
  • No. of Keyed Items
    No. of Parts
    Width
    Length
    Height
    Availability
    Price
  • 470014-418
    Frog, Female
    Frog, Female
    45
    1
    15 cm
    36 cm
    6 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,000.00
    $2,000.00
    Each (1UN)
       $2,000.00
  • 470014-416
    Frog, Male
    Frog, Male
    48
    1
    15 cm
    33 cm
    6 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,130.00
    $2,130.00
    Each (1UN)
       $2,130.00