Friction Rods
Supplier: LAIRD PLASTICS INC CP
Jump to:
About this item
For your Electrostatic Needs!
- Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
4 Options Available Below
Product Details & Documents
For your Electrostatic Needs!
- Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Product Family Options
Product Information
- AvailabilityPrice
- 470005-504Solid Acrylic, Flat EndedSupplier Number: PER QUOTE
- 470013-466Hollow Glass, RoundedSupplier Number: FRHG12
- 470016-220Solid Glass, RoundedSupplier Number: FRSG12
- 470104-254Hard Nylon, Flat EndedSupplier Number: BLNYLROD
- 470230-306Ebonite RodEbonite RodDiscontinued-Supplier Number: PH0856A