Friction Rod Demonstration
Friction Rod Demonstration
Catalog # 470005-502
Supplier: Avantor
Jump to:
Product Details & Documents
Perfect for Demonstrating Electrostatics
The set comes with everything you need to create and dissipate static charges, including a hollow glass friction rod, hard rubber rod, three friction pads (silk, flannel, and animal fur), nylon string, balloons, solid plastic rod, and instructions.
Specifications
- Description:Friction Rod Demonstration