Friction Block Set
Friction Block Set
Catalog # 470149-098
Supplier: VWR International
Jump to:
Product Details & Documents
The Friction Block Set is ideal for simple friction experiments.
- Ideal for Simple Friction Experiments
- Instructions Included
- Attached Mirror
This set consists of a glass mirror with sandpaper affixed to one side and two wooden blocks: one with a hook and the other with an eye. The latter block also has a mirror cemented to one face.
It comes with instructions for use.
Especificaciones
- Description:Friction Block Set