Four Stroke Diesel Engine
Four Stroke Diesel Engine
470014-348
Ideal for Individual Study
Compact enough to fit on a desktop, yet with all the features of a full size diesel engine, this is an ideal demonstration for the classroom. Functional components of this after cooled diesel engine model include camshaft, rocks-arms, and tappets „ all clearly demonstrated. This chain driven overhead camshaft type engine also features a functioning fuel injection system. Ignition is shown by means of a miniature bulb. Size: Size: 8 1/4" x 8 1/4" x 14 1/2".
- Description:Four Stroke Diesel Engine