Forensic Science: Introduction to DNA Fingerprinting Kit
Catalog # 470017-042
Supplier: LAB-AIDS INC (KITS) SE
Specifications
- Descripción:Forensic Science: Introduction to DNA Fingerprinting Kit
- Referencia:470017-042
- Nº de proveedor:1271
Specifications
About this item
Gives students a real feel for what it takes to be a real detective. This basic introduction to forensic science covers many bases, including cells, cell structure, and magnification.
- This kit contains five activities and material for 15 groups of two students
- Material is reusable
Students use this information to construct a model 'chromosome' with color-coded 'genes' representing some physical traits of the suspect. The process of DNA mapping and DNA fingerprinting is carefully explained. Students will be able to read simulated DNA x-ray films to positively identify the perpetrators of the crime.