Orders LinkContactUsLinkComponent
United Scientific™ Push/Pull Spring Scales
United Scientific™ Push/Pull Spring Scales
Certificates
United Scientific™ Push/Pull Spring Scales
United Scientific™ Push/Pull Spring Scales
6 Options Available Below
Product Details & Documents
Versatile spring scale.
  • Weighs in Grams and Newtons
  • Color coded to differentiate capacity
These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.
 : Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.
 : This product is for ages six and up.
Product Family Options
Información del producto
  • Weighing Capacity
    Availability
    Price
  • 470150-550
    Push/pull spring scale, blue
    Push/pull spring scale, blue
    250 g/2.5 N, graduation interval: 0.05 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $140.20
    $140.20
    Each (1UN)
       $140.20
    Supplier Number: PS0250
  • 470017-446
    Push/pull spring scale, green
    Push/pull spring scale, green
    500 g/5 N, graduation interval: 0.1 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $140.20
    $140.20
    Each (1UN)
       $140.20
    Supplier Number: PS0500
  • 470150-486
    Push/pull spring scale, brown
    Push/pull spring scale, brown
    1000 g/10 N, graduation interval: 0.2 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $142.53
    $142.53
    Log in Para disponibilidad
    Case de 150 (1UN)
    Case de 150 (1UN) - $155.36
    $155.36
    Each (1UN)
       $142.53
    Case de 150 (1UN)
       $155.36
    Supplier Number: PS1000
  • 470150-474
    Push/pull spring scale, red
    Push/pull spring scale, red
    2000 g/20 N, graduation interval: 0.4 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $140.20
    $140.20
    Each (1UN)
       $140.20
    Supplier Number: PS2000
  • 470150-868
    Push/pull spring scale, cream
    Push/pull spring scale, cream
    3000 g/30 N, graduation interval: 0.5 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $140.20
    $140.20
    Each (1UN)
       $140.20
    Supplier Number: PS3000
  • 470150-620
    Push/pull spring scale, yellow
    Push/pull spring scale, yellow
    5000 g/50 N, graduation interval: 1 N
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $140.20
    $140.20
    Each (1UN)
       $140.20
    Supplier Number: PS5000