Flint Glass Watch Glasses
Proveedor: UNITED SCIENTIFIC SUPPLIES
Certificados
Acerca de este artículo
Useful clear watch glasses for evaporation.
- Flint glass
- Flame polished edges
Made from clear flint glass (1.8 to 2.0mm thick), fine ground and polished edges.
Frequently Bought Together