Flat Plastic Meter Stick
Flat metric meter stick, Plastic, 1 m
Catalog # 470106-454
Supplier: SPECTRUM EDUCATIONAL SUPPLIES
Supplier #: 11050-B
Jump to:
Product Details & Documents
Flexible plastic meter stick marked in cm with 1 mm divisions on one side and with alternate-colored dm bars on the other, to show the relationships between units.
Flexible plastic meter stick marked in cm with 1 mm divisions on one side and with alternate-colored dm bars on the other, to show the relationships between units.
Especificaciones
- Description:Flat metric meter stick
- Length:1 m
- Material:Plastic