Flask Brushes
Supplier: ALLIED K & R
Jump to:
Acerca de este artículo
White bristle and plastic handle.
- Multiple sizes available
Recommended for volumetric and boiling flasks because of the unusally flexible, curved steel spring that enables the brush to reach all parts of the flask.
3 Options Available Below
Product Details & Documents
White bristle and plastic handle.
- Multiple sizes available
Recommended for volumetric and boiling flasks because of the unusally flexible, curved steel spring that enables the brush to reach all parts of the flask.
Product Family Options
Información del producto
- Bristle ColorHandle MaterialTotal lengthAvailabilityPrice
- WhitePlastic12"Each (1UN)$125.22Case de 144 (1UN)$136.49Supplier Number: D130
- Supplier Number: D131
- WhitePlastic17"Each (1UN)$148.24Case de 144 (1UN)$161.59Supplier Number: D132