Orders LinkContactUsLinkComponent
Ward's® Fixed 90 ̊ Photo Rules
Ward's® Fixed 90 ̊ Photo Rules
Ward's® Fixed 90 ̊  Photo Rules
Ward's® Fixed 90 ̊ Photo Rules
Easy to Measure at Right Angles
  • Easy to Read Photo Rules
  • Both English and Metric Scales
Precisely measure objects in two dimensions with these one-sided, "L-shaped", durable plastic rules. The English rule is 4" per side and is graduated in ¹⁄₁₆" increments; the metric rule is 11 cm per side and is graduated in 1 mm increments.
Product Family Options
  • Description
    Width
    Height
  • Fixed 90° photo rule, imperial, ¹⁄₁₆" increments
    4"
    4"
    Each (1UN)
    Each (1UN) - $117.28
    $117.28
    Each (1UN)
       $117.28
     : 4CSFWH3X3PLPGWARDS
  • Fixed 90° photo rule, metric
    11 cm
    11 cm
    Each (1UN)
    Each (1UN) - $117.28
    $117.28
    Each (1UN)
       $117.28
     : 4CSM-WH8X8PL/WARDS