Fingertip Safety Tape
Safety Tape, Finger, 27 m long x 1.9 cm wide
Catalog # 470150-976
Supplier: GENERAL BANDAGES INC.
Número de proveedor #: 41408-3/4
Jump to:
Product Details & Documents
Protect your fingers without gloves.
This self-adhering tape maintains an unimpaired sense of touch, yet prevents hand and finger injuries from sharp edges, hot surfaces, or rough materials.
Made from pure surgical grade cotton gauze coated with natural latex rubber. Roll measures 1.9 cm wide × 27 m long.
Especificaciones
- Description:Safety Tape, Finger, 27 m long x 1.9 cm wide