Fine-Tip Brushes
Fine-Tip Brushes, Camel hair, 17.8 cm (7")
Catalog # 470020-022
Supplier: SOLO HORTON BRUSHES, INC.
Número de proveedor #: 2115-2
Jump to:
Product Details & Documents
The soft, natural camel hair bristles of this fine tipped brush will not harm delicate structures like stiff forceps or needles will.
Especificaciones
- Bristle Material:Camel hair
- Description:Fine-Tip Brushes
- Total length:17.8 cm (7")