Orders LinkContactUsLinkComponent
Field Guide for Soil and Stratigraphic Analysis Chart
Field Guide for Soil and Stratigraphic Analysis Chart
Field Guide for Soil & Straigraphic Analysis Chart
Catalog # 470018-588
Número de proveedor #:  FG SOIL
Field Guide for Soil and Stratigraphic Analysis Chart
Catalog # 470018-588
Field Guide for Soil and Stratigraphic Analysis Chart
Field Guide for Soil and Stratigraphic Analysis Chart
Field Guide for Soil & Straigraphic Analysis Chart
Price
$532.49
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Enhance Your Soil and Sediment Descriptions
Improve the accuracy of your boring logs and classification of soils in the field using this field guide. The front side of the text includes a flow chart listing key descriptive elements necessary for characterizing sedimentary sequences on boring logs. Common terms from geologic, engineering, and soil science disciplines are also used to help prepare meaningful boring logs that are clear and concise. The back side includes an easy-to-use flow chart that takes the mystery out of using the Unified Soil Classification System and assigning a group name and USCS symbol in the field. Guide is printed on durable, double-sided, plastic-coated paper. Size: 8 1/2" x 11".
Especificaciones
  • Description:
    Field Guide for Soil & Straigraphic Analysis Chart