Fiber Optic Holder for Carousel Power Supply
Catalog # 470018-232
Supplier: STEINER ENTERPRISES SE
Specifications
- Descripción:Fiber Optic Holder for Carousel Power Supply
- Referencia:470018-232
- Nº de proveedor:SE1041-00
Specifications
About this item
Dual purpose for Dual Benefits
- The Fiber Optics Holder is an accessory for the Carousel Spectrum Tube Power Supply with two great features
The Fiber Optics Holder holds the end of the fiber optic probe of a digital spectrometer in place when in front of the illuminated spectrum tube in the Carousel Power Supply, allowing the probe to be positioned optimally to receive the maximum amount of light from the tube. It also holds the probe end at a consistent safe distance from the tube surface.