Fever Tester
Fever Tester
Catalog # 470146-260
Supplier: LIQUID CRYSTAL RESOURCES SE
Número de proveedor #: BFT45
Jump to:
Product Details & Documents
Accurate Readings in Fifteen Seconds
Simply press this liquid crystal thermometer against the forehead and in 15 seconds the correct temperature is highlighted in green. No sterilization is required. Range: 95–104°F. Length: 3 1/2".
Especificaciones
- Description:Fever Tester