Fermentation Tubes
Fermentation Tube, Ungraduated, 5 mL, with Foot
Catalog # 470235-690
Supplier: GSC INTERNATIONAL SE
Supplier #: 601-9-10
Jump to:
Product Details & Documents
Verify gas production in fermentation exercises, gas produced by yeast or other organisms rises into a 5 or 10 ml evolution tube.
- Ungraduated with foot
Specifications
- Description:Fermentation Tube, Ungraduated, 5 mL, with Foot