Orders LinkContactUsLinkComponent
Fast Release Pi-Pumps
Supplier:   BEL-ART PRODUCTS SE
Fast Release Pi-Pumps
Certificates
Fast Release Pi-Pumps
Fast Release Pi-Pumps
2 Options Available Below
Product Details & Documents
Designed for fast and accurate one-handed operation.
  • Reduce the risk of injury from repetitive pipetting
  • Expel entire contents in one shot or smaller amounts precisely
Each pump is resistant to acids, bases, and most solvents; and it can be easily disassembled for cleaning.
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 470153-804
    Fast Release Pi-Pump, Green
    Fast Release Pi-Pump, Green
    10 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $976.00
    $976.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,372.56
    $1,372.56
    Each (1UN)
       $976.00
    Case de 12 (1UN)
       $1,372.56
    Supplier Number: F37904-0010
  • 470153-802
    Fast Release Pi-Pump, Red
    Fast Release Pi-Pump, Red
    25 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,170.00
    $1,170.00
    Log in Para disponibilidad
    Case de 12 (1UN)
    Case de 12 (1UN) - $1,645.00
    $1,645.00
    Each (1UN)
       $1,170.00
    Case de 12 (1UN)
       $1,645.00
    Supplier Number: F37904-0025