Eyepieces
Supplier: MOTIC HONG KONG LIMITED
Jump to:
Acerca de este artículo
Eyepieces with or without cross hairs for standard monocular microscope.
2 Options Available Below
Product Details & Documents
Eyepieces with or without cross hairs for standard monocular microscope.
Product Family Options
Información del producto
- AvailabilityPrice
- 470014-566Eyepiece, DIN, 10× WF, w/Cross HairsSupplier Number: MA-EF-1M1O
- 470103-530Eyepiece, DIN, 10× WFSupplier Number: MA-EF1W10