Extended Reach Screwdriver Set
Screwdriver, Set, 5 Piece Extended Reach
Catalog # 470004-750
Supplier: MOODY TOOLS INC SE
Número de proveedor #: 58-0460
Jump to:
Product Details & Documents
These set of extended reach screwdrivers (.093”, .125”. .141” phillips #00, phillips #0), is perfect for delicate microscope repair/adjustment. Blades are manufactured from hardened, high tensile strength, alloy steel, securely molded into the ergonomic, anti-static handle. Made in the U.S.A., and backed by a lifetime warranty.
Delivery information: Supplied in a durable, vinyl pouch.
Especificaciones
- Description:Screwdriver, Set, 5 Piece Extended Reach