Ward's® Examining Forensic Science Poster
Catalog # 470023-262
Supplier: Avantor
Specifications
- Descripción:WARD’S Examining Forensic Science Poster
- Referencia:470023-262
- Nº de proveedor:WARD470023-262
Specifications
About this item
Demonstrate Real World Applications of Conventional Science
- Size: 21 x 34"
Introduce students to several branches of forensic science with this visually stimulating poster. Topics discussed include Odontology, Serology, Pathology, Firearms, Anthropology, Trace evidence, and Chemistry/toxicology. Size: 21" x 34".