Euplotes Slide
Euplotes, Whole mount, Fast Green and Mallory's
Catalog # 470181-466
Supplier: Avantor
Jump to:
Product Details & Documents
Single Celled Flagellates
- Identifying features clearly distinguishable
- Rigorous quality control standards
A hypotrich having fused cilia or cirri.
Especificaciones
- Description:Euplotes
- Stain:Fast Green and Mallory's
- Orientation:Whole mount