Orders LinkContactUsLinkComponent
Eudiometer
Supplier:   VWR International
Eudiometer
Supplier:  VWR International
Certificates
Eudiometer
2 Options Available Below
Product Details & Documents
Demonstrate the combining volume ratios of gases.
  • Two sizes available
  • Platinum electrodes included
Long, sturdy glass tubes have platinum electrodes sealed at the closed end with a 2 mm arc gap. The tubes are marked with a permanent brown scale in 0.1 or 0.2 mL divisions for 50 and 100 mL respectively. An induction coil is recommended for use with eudiometers.
Product Family Options
Información del producto
  • Availability
    Price
  • 470219-674
    Eudiometer, 50 ml
    Eudiometer, 50 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $621.28
    $621.28
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $677.19
    $677.19
    Log in Para disponibilidad
    Case de 48 (1UN)
    Case de 48 (1UN) - $29,821.35
    $29,821.35
    Each (1UN)
       $621.28
    Paquete de 12 (1UN)
       $677.19
    Case de 48 (1UN)
       $29,821.35
  • 470235-712
    Eudiometer, 100 ml
    Eudiometer, 100 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,008.80
    $1,008.80
    Log in Para disponibilidad
    Case de 10 (1UN)
    Case de 10 (1UN) - $1,099.59
    $1,099.59
    Each (1UN)
       $1,008.80
    Case de 10 (1UN)
       $1,099.59