ContactUsLinkComponent
Estwing Polished Pick-Head Hammer, 22 oz.
Estwing Polished Pick-Head Hammer, 22 oz.
  470199-828
undefined
Estwing Polished Pick-Head Hammer, 22 oz.
  470199-828
 :  E3-22P
  • Descripción:
    Estwing Polished Pick-Head Hammer, 22 oz, Nylon-Vinyl Grip
  • Referencia:
    470199-828
  • Nº de proveedor:
    E3-22P
Versatile Enough for All Hard-Rock Applications
  • Standard pick head rock hammer
  • Durable construction and comfortable grip
Long regarded as the finest in lapidary and field tools, Estwing hammers have a strong, solid steel I-beam construction that virtually eliminates the risks of the head loosening or the handle breaking as might occur with other models.
The grip provides deep, comfortable cushioning that makes the tool easier to hold and absorbs more shock, which is especially valuable to those who use hammers for extended periods of time.
Nylon-vinyl cushion grip. Length: 13".