Equisetum, Aerials Stem Slide
Equisetum, Aerials Stem, Cross section, Quadruple
Catalog # 470181-150
Supplier: Avantor
Jump to:
Product Details & Documents
Vascular Plant that Reproduces by Spores and not Seeds
- Identifying features clearly distinguishable
- Rigorous quality control standards
Demonstrates an ectophloic siphonostele.
Especificaciones
- Description:Equisetum, Aerials Stem
- Stain:Quadruple
- Orientation:Cross section