Equilateral Refraction Prism
Equilateral Refraction Prism
Catalog # 470021-796
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: FGP075
Jump to:
Product Details & Documents
Demonstrate the bending properties of light.
- Clear flint glass
- Demonstrate refraction and reflection
This flint glass prism measures 3" long by 3/8" thick.
Specifications
- Description:Equilateral Refraction Prism