Equilateral Prisms
Prism, Equilateral, 25 mm x 50 mm
Catalog # 470003-426
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: PGE050
Jump to:
Product Details & Documents
Easily demonstrate the separation of light.
- Demonstrate reflection and refraction
- Flint glass
Excellent quality, polished flint glass prisms for experiments in diffraction, dispersion, and spectrum formation. Individually boxed to prevent chipping.
Specifications
- Description:Prism, Equilateral, 25 mm x 50 mm