Equally Tempered Tuning Forks Set
Acerca de este artículo
Both of these sets come with a roll-up style canvas storage pouch and a solid rubber activator.
- Set of 8 or 13 tuning forks
- Storage pouch and activator included
The set of eight contains the following frequencies: 261.6, 293.7, 329.6, 349.2, 392.0, 440, 493.9 and 523.2. The set of 13 contains the following frequencies: 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392.0, 415.3, 440, 466.1, 493.9 and 523.2.
2 Options Available Below
Product Details & Documents
Both of these sets come with a roll-up style canvas storage pouch and a solid rubber activator.
- Set of 8 or 13 tuning forks
- Storage pouch and activator included
The set of eight contains the following frequencies: 261.6, 293.7, 329.6, 349.2, 392.0, 440, 493.9 and 523.2. The set of 13 contains the following frequencies: 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392.0, 415.3, 440, 466.1, 493.9 and 523.2.
Product Family Options
Información del producto
- AvailabilityPrice
- 470148-824Tuning fork, equally tempered, set/8Tuning fork, equally tempered, set/8Kit de 8$39.40Case de 20 (8UN)$788.00
- 470104-588Tuning fork, equally tempered, set/13, machine made from aluminum alloyTuning fork, equally tempered, set/13, machine made from aluminum alloyLog in Para disponibilidadKit de 1$50.15