Equal Volume Density Set, Rectangular
Catalog # 470149-614
Supplier: EISCO SCIENTIFIC LLC
Specifications
- Descripción:Equal volume density set, set of 4
- Referencia:470149-614
- Nº de proveedor:PH0111
Specifications
About this item
Rectangular density set.
- Set of 4 equal volume rectangular prisms
- Includes: Aluminum, Copper, Steel and Brass
- Dimensions: 1.25×1.25×5 cm (0.5×0.5×2")
- Machine tooled to the most exacting tolerances in the industry
This 4 piece set includes aluminum, brass, copper and steel specimens of equal volume designed for successful density experiments and specific gravity measurement.