Epiglottis Slide
Epiglottis, Longitudinal section, Hematoxylin and Eosin
Catalog # 470182-894
Supplier: Avantor
Jump to:
Product Details & Documents
Located at the Base of the Tongue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stratified squamous epithelium, cartilage, and surrounding tissues.
Specifications
- Description:Epiglottis
- Stain:Hematoxylin and Eosin
- Orientation:Longitudinal section