Energy Conversion Demonstrator
Energy Conversion Demonstrator
470002-940
: ECK001
Demonstrate the conversion energy into different forms.
- All have easy to connect banana plugs
- Demonstrate chemical, light, wind and mechanical energy
Illustrate the changing of energy from one form to another with this classroom demonstration. Three energy sources are provided mounted on a board– a battery, solar cell, and DC generator — for a series of experiments and energy conversions Sound, light, heat, magnetic, and mechanical transducers are also included mounted on accessory boards. Includes an instruction manual. Size: 35 x 35.5 x 12.7cm.
- Description:Energy Conversion Demonstrator