Endocrine System Slide Set
Endocrine System Slide Set, 4, Animal
Catalog # 470183-598
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Gland Types that Secrete Directly into the Blood
- Identifying features clearly distinguishable
- Rigorous quality control standards
All are sections.
Ordering information: Slide Set contents include Adrenal, Pituitary, Thyroid, Thyroid and Parathyroid.
Documents
Endocrine System Slide Set
Documentación del producto
Especificaciones
- Type:Animal
- Description:Endocrine System Slide Set
- No. of Slides:4