Orders LinkContactUsLinkComponent
Enameled Copper Magnet Wire
Enameled Copper Magnet Wire
Enameled Copper Magnet Wire
Certificates
Enameled Copper Magnet Wire
3 Options Available Below
Product Details & Documents
A high quality enameled, copper magnet wire.
The 454 g spools are available in several standard AWG sizes.
Product Family Options
Información del producto
  • Availability
    Price
  • 470014-912
    Wire, Enameled Copper Magnet, 18 Gauge, 61 m, 454 grams
    Wire, Enameled Copper Magnet, 18 Gauge, 61 m, 454 grams
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $734.49
    $734.49
    Each (1UN)
       $734.49
    Supplier Number: E18
  • 470149-670
    Wire, Enameled Copper Magnet, 20 Gauge, 94 m, 454 grams
    Wire, Enameled Copper Magnet, 20 Gauge, 94 m, 454 grams
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $734.49
    $734.49
    Each (1UN)
       $734.49
    Supplier Number: E-20
  • 470104-612
    Wire, Enameled Copper Magnet, 22 Gauge, 149 m, 454 grams
    Wire, Enameled Copper Magnet, 22 Gauge, 149 m, 454 grams
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $763.26
    $763.26
    Each (1UN)
       $763.26
    Supplier Number: E-22