Elodea, Stem Slide
Elodea, Stem, Cross section, Quadruple
470181-236
: Avantor
Illustrates Aquatic Plant Adaptations
- Identifying features clearly distinguishable
- Rigorous quality control standards
Waterweed. A simple hydrophyte.
- Description:Elodea, Stem
- Stain:Quadruple
- Orientation:Cross section