Orders LinkContactUsLinkComponent
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Certificates
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit explores chemical nomenclature through experimentation.
  • Ideal for Grades 7-10
  • Students Learn to Recognize Evidence of Chemical Changes
  • Kit Illustrates Concepts of Elements, Compounds, and Mixtures
  • Special Emphasis on Creating New Compounds in Chemical Reactions
  • Enough Material to Repeat Experiment Twice
This kit illustrates the concepts of elements, compounds, and mixtures with special emphasis on creating new compounds in chemical reactions. Students witness evidence of chemical changes identified by the formation of precipitates, color changes, odors, or the production of gas.
The kit includes enough material to repeat the experiment twice. Worksheets focus on chemical symbols, chemical formulas, chemical names, components of common compounds, and distinguishing between chemical and physical changes.
Ordering information: This lab includes an instructor manual, a SDS, filter paper, spoons, calcium chloride, sodium carbonate, copper sulfate, and sodium bicarbonate. Balance and laboratory supplies are needed but not included.
Product Family Options
Product Information
  • Includes
    No. of Lab Groups
    Availability
    Price
  • 470200-078
    Elements, Compounds, and Mixtures II Lab Activity
    Elements, Compounds, and Mixtures II Lab Activity
    Instructions and Student Worksheets
    6
    Discontinued
    Supplier Number: 13-108
  • 470002-478
    Elements, Compounds, and Mixtures II Lab Activity
    Elements, Compounds, and Mixtures II Lab Activity
    6
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,837.65
    $3,837.65
    Each (1UN)
       $3,837.65
    Supplier Number: 13-108
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 